Merck
  • Grooved nanoplate assembly for rapid detection of surface enhanced Raman scattering.
Grooved nanoplate assembly for rapid detection of surface enhanced Raman scattering.
Nanoscale (2017-10-05)
Xuan Liu, Dan Wu, Qing Chang, Jing Zhou, Yongyi Zhang, Zhaona Wang
ABSTRACT
Rapid detection of surface enhanced Raman scattering (SERS) signals is in great demand in the fields of biological medicine and environmental monitoring. Herein, a grooved silver nanoplate assembly (GSNA) with an abundance of multiscale gaps has been proposed for the first time and skillfully synthesized to act as an excellent platform for surface enhanced Raman spectroscopy with ultrafast and ultrasensitive detection. By effectively combining the hotspots effect of nanogaps and the trapping effect of gaps in the scale of subwavelength, the Raman signal was greatly enhanced by a factor of 10
Sigma-Aldrich
Silver nitrate, 99.9999% trace metals basis