Merck
  • Dose-Specific Effects of Di-Isononyl Phthalate on the Endocannabinoid System and on Liver of Female Zebrafish.
Dose-Specific Effects of Di-Isononyl Phthalate on the Endocannabinoid System and on Liver of Female Zebrafish.
Endocrinology (2017-09-25)
Isabel Forner-Piquer, Francesca Maradonna, Giorgia Gioacchini, Stefania Santangeli, Marco Allarà, Fabiana Piscitelli, Hamid R Habibi, Vincenzo Di Marzo, Oliana Carnevali
ABSTRACT
Phthalates, used as plasticizers, have become a ubiquitous contaminant and have been reported for their potential to induce toxicity in living organisms. Among them, di-isononyl phthalate (DiNP) has been recently used to replace di(2-ethylhexyl) phthalate (DEHP). Nowadays, there is evidence that DiNP is an endocrine-disrupting chemical; however, little is known about its effects on the endocannabinoid system (ECS) and lipid metabolism. Hence, the aim of our study was to investigate the effects of DiNP on the ECS in zebrafish liver and brain and on hepatic lipid storage. To do so, adult female zebrafish were exposed to three concentrations (0.42 µg/L, 4.2 µg/L, and 42 µg/L) of DiNP via water for 3 weeks. Afterwards, we investigated transcript levels for genes involved in the ECS of the brain and liver as well as liver histology and image analysis, Fourier-transform infrared spectroscopy imaging, and measurement of endocannabinoid levels. Our results demonstrate that DiNP upregulates orexigenic signals and causes hepatosteatosis together with deregulation of the peripheral ECS and lipid metabolism. A decrease in the levels of ECS components at the central level was observed after exposure to the highest DiNP concentration tested. These findings suggest that replacement of DEHP with DiNP should be considered with caution because of observed adverse DiNP effects on aquatic organisms.
Sigma-Aldrich
Ethyl 3-aminobenzoate, 97%
Sigma-Aldrich
Arachidonylethanolamide, ≥97.0% (TLC), oil
Sigma-Aldrich
N-Oleoylethanolamine, ~98% (TLC)
Sigma-Aldrich
Palmitoylethanolamide