Merck
  • Nitric Oxide Mediates Nitrite-Sensing and Acclimation and Triggers a Remodeling of Lipids.
Nitric Oxide Mediates Nitrite-Sensing and Acclimation and Triggers a Remodeling of Lipids.
Plant physiology (2017-09-20)
Lina-Juana Dolch, Josselin Lupette, Guillaume Tourcier, Mariette Bedhomme, Séverine Collin, Leonardo Magneschi, Melissa Conte, Khawla Seddiki, Christelle Richard, Erwan Corre, Laurent Fourage, Frédéric Laeuffer, Robert Richards, Michael Reith, Fabrice Rébeillé, Juliette Jouhet, Patrick McGinn, Eric Maréchal
ABSTRACT
Nitric oxide (NO) is an intermediate of the nitrogen cycle, an industrial pollutant, and a marker of climate change. NO also acts as a gaseous transmitter in a variety of biological processes. The impact of environmental NO needs to be addressed. In diatoms, a dominant phylum in phytoplankton, NO was reported to mediate programmed cell death in response to diatom-derived polyunsaturated aldehydes. Here, using the
Sigma-Aldrich
S-Nitroso-N-acetyl-DL-penicillamine, ≥97%, powder
Sigma-Aldrich
1,2-Dioleoyl-sn-glycerol, ≥97%