Merck
  • S100B promotes injury-induced vascular remodeling through modulating smooth muscle phenotype.
S100B promotes injury-induced vascular remodeling through modulating smooth muscle phenotype.
Biochimica et biophysica acta (2017-07-12)
Teng Cao, Lei Zhang, Ling-Ling Yao, Fei Zheng, Lu Wang, Jian-Ye Yang, Ling-Yun Guo, Xing-Yuan Li, Yu-Wen Yan, Ya-Mu Pan, Miao Jiang, Long Chen, Jun-Ming Tang, Shi-You Chen, Jia-Ning Wang
ABSTRACT
S100B is a biomarker of nervous system injury, but it is unknown if it is also involved in vascular injury. In the present study, we investigated S100B function in vascular remodeling following injury. Balloon injury in rat carotid artery progressively induced neointima formation while increasing S100B expression in both neointimal vascular smooth muscle (VSMC) and serum along with an induction of proliferating cell nuclear antigen (PCNA). Knockdown of S100B by its shRNA delivered by adenoviral transduction attenuated the PCNA expression and neointimal hyperplasia in vivo and suppressed PDGF-BB-induced VSMC proliferation and migration in vitro. Conversely, overexpression of S100B promoted VSMC proliferation and migration. Mechanistically, S100B altered VSMC phenotype by decreasing the contractile protein expression, which appeared to be mediated by NF-κB activity. S100B induced NF-κB-p65 gene transcription, protein expression and nuclear translocation. Blockade of NF-κB activity by its inhibitor reversed S100B-mediated downregulation of VSMC contractile protein and increase in VSMC proliferation and migration. It appeared that S100B regulated NF-κB expression through, at least partially, the Receptor for Advanced Glycation End products (RAGE) because RAGE inhibitor attenuated S100B-mediated NF-κB promoter activity as well as VSMC proliferation. Most importantly, S100B secreted from VSMC impaired endothelial tube formation in vitro, and knockdown of S100B promoted re-endothelialization of injury-denuded arteries in vivo. These data indicated that S100B is a novel regulator for vascular remodeling following injury and may serve as a potential biomarker for vascular damage or drug target for treating proliferative vascular diseases.
Sigma-Aldrich
MISSION® esiRNA, targeting human S100B
Pricing and availability is not currently available.