Merck
  • Pediatric Cytochrome P450 Activity Alterations in Nonalcoholic Steatohepatitis.
Pediatric Cytochrome P450 Activity Alterations in Nonalcoholic Steatohepatitis.
Drug metabolism and disposition: the biological fate of chemicals (2017-10-08)
Hui Li, Mark J Canet, John D Clarke, Dean Billheimer, Stavra A Xanthakos, Joel E Lavine, Robert P Erickson, Nathan J Cherrington
ABSTRACT
Variable drug responses depend on individual variation in the activity of drug-metabolizing enzymes, including cytochrome P450 enzymes (CYP). As the most common chronic liver disease in children and adults, nonalcoholic steatohepatitis (NASH) has been identified as a source of significant interindividual variation in hepatic drug metabolism. Compared with adults, children present age-related differences in pharmacokinetics and pharmacodynamics. The purpose of this study was to determine the impact of fatty liver disease severity on the activity of a variety of CYP enzymes in children and adolescents. Healthy and nonalcoholic fatty liver disease pediatric subjects aged 12-21 years inclusive received an oral cocktail of four probe drugs: caffeine (CYP1A2, 100 mg), omeprazole (CYP2C19, 20 mg), losartan (CYP2C9, 25 mg), and midazolam (CYP3A4, 2 mg). Venous blood and urine were collected before administration and 1, 2, 4, and 6 hours after administration. Concentrations of the parent drugs and CYP-specific metabolites were quantified in plasma and urine using liquid chromatography with tandem mass spectrometry. In plasma, the decreased metabolic area under the curve (AUC) ratio, defined as the metabolite AUC to parent AUC, of omeprazole indicated significant decreases of CYP2C19 (
Sigma-Aldrich
1,7-Dimethylxanthine, ~98%, solid
Supelco
5-Hydroxyomeprazole, analytical standard
Sigma-Aldrich
1′-Hydroxymidazolam