Merck
  • Hedgehog/Gli1 signaling pathway regulates MGMT expression and chemoresistance to temozolomide in human glioblastoma.
Hedgehog/Gli1 signaling pathway regulates MGMT expression and chemoresistance to temozolomide in human glioblastoma.
Cancer cell international (2017-12-12)
Ke Wang, Dongjiang Chen, Zhouqi Qian, Daming Cui, Liang Gao, Meiqing Lou
ABSTRACT
Chemoresistance of glioblastoma (GBM) is a feature of this devastating disease. This study is to determine the relationship between Hedgehog (HH)/Gli1 signaling pathway and chemoresistance to temozolomide (TMZ) in human GBM. We analyzed Gli1 nuclear staining and O O This study shows that HH/Gli1 signaling pathway regulates MGMT expression and chemoresistance to TMZ in human GBM independent from MGMT promoter methylation status, which offers a potential target to restore chemosensitivity to TMZ in a fraction of GBM with high MGMT expression.
Sigma-Aldrich
Sodium bisulfite solution, purum, ~40%