Merck
TIM-3 Regulates CD103
Cancer cell (2018-01-10)
Álvaro de Mingo Pulido, Alycia Gardner, Shandi Hiebler, Hatem Soliman, Hope S Rugo, Matthew F Krummel, Lisa M Coussens, Brian Ruffell
ABSTRACT
Intratumoral CD103