Merck
Leukemia-specific delivery of mutant NOTCH1 targeted therapy.
The Journal of experimental medicine (2017-11-22)
Giovanni Roti, Jun Qi, Samuel Kitara, Marta Sanchez-Martin, Amy Saur Conway, Anthony C Varca, Angela Su, Lei Wu, Andrew L Kung, Adolfo A Ferrando, James E Bradner, Kimberly Stegmaier
ABSTRACT
On-target drug delivery remains a challenge in cancer precision medicine; it is difficult to deliver a targeted therapy to cancer cells without incurring toxicity to normal tissues. The SERCA (sarco-endoplasmic reticulum Ca
Sigma-Aldrich
Filipin complex from Streptomyces filipinensis, ≥70% (UV)