Merck
  • A Fluorescence-Based High-Throughput Assay for the Identification of Anticancer Reagents Targeting Fructose-1,6-Bisphosphate Aldolase.
A Fluorescence-Based High-Throughput Assay for the Identification of Anticancer Reagents Targeting Fructose-1,6-Bisphosphate Aldolase.
SLAS discovery : advancing life sciences R & D (2017-08-19)
Eun Jeong Cho, Ashwini K Devkota, Gabriel Stancu, Ramakrishna Edupunganti, Garth Powis, Kevin N Dalby
ABSTRACT
A high rate of glycolysis, which supplies energy and materials for anabolism, is observed in a wide range of tumor cells, making it a potential pathway to control cancer growth. ALDOA is a multifunctional enzyme in the glycolytic pathway and also promotes HIF-1α, which is of importance in hypoxic solid tumors. The current method for assaying ALDOA activity involves monitoring the consumption of NADH in vitro using absorbance or intrinsic fluorescence via a coupled enzymatic reaction. Here, we report the development of a homogeneous biochemical assay that can overcome limitations of current methods, in particular for the application of high-throughput drug screening. The assay utilizes the commercially available Elite NADH Assay Kit, which incorporates an enzymatic reaction to measure the level of NADH using a fluorescent probe. Assay optimization and validation are discussed. Its feasibility for high-throughput screening (HTS) was demonstrated by screening 65,000 compounds for the identification of small molecules that inhibit ALDOA. Through a validation screen and dose-response evaluation, four inhibitors with IC50 below 10 µM were identified. In conclusion, we demonstrate that a traditional ALDOA assay can be transformed readily into a fluorescence-based assay utilizing a commercial NADH detection kit that is rapid, sensitive, inexpensive, and HTS friendly.
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
α-Glycerophosphate Dehydrogenase-Triosephosphate Isomerase from rabbit muscle, Type III, ammonium sulfate suspension
Sigma-Aldrich
Phosphate buffered saline, tablet
Sigma-Aldrich
ECO TWEEN® 20, viscous liquid
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
Bovine Serum Albumin, lyophilized powder, ≥96% (agarose gel electrophoresis)
Sigma-Aldrich
α-Glycerophosphate Dehydrogenase from rabbit muscle, Type X, lyophilized powder, ≥100 units/mg protein
Sigma-Aldrich
TWEEN® 20, viscous liquid