Merck
  • Functions of maize genes encoding pyruvate phosphate dikinase in developing endosperm.
Functions of maize genes encoding pyruvate phosphate dikinase in developing endosperm.
Proceedings of the National Academy of Sciences of the United States of America (2017-12-20)
Ryan R Lappe, John W Baier, Susan K Boehlein, Ryan Huffman, Qiaohui Lin, Fabrice Wattebled, A Mark Settles, L Curtis Hannah, Ljudmilla Borisjuk, Hardy Rolletschek, Jon D Stewart, M Paul Scott, Tracie A Hennen-Bierwagen, Alan M Myers
ABSTRACT
Maize