Merck
  • Biotransformation of Progesterone by the Ascomycete Aspergillus niger N402.
Biotransformation of Progesterone by the Ascomycete Aspergillus niger N402.
Biochemistry. Biokhimiia (2018-03-15)
O S Savinova, P N Solyev, D V Vasina, T V Tyazhelova, T V Fedorova, T S Savinova
ABSTRACT
The ability of the ascomycete Aspergillus niger N402 to transform exogenous progesterone was investigated. We found that this strain has steroid-hydroxylating activity and can introduce a hydroxyl group into the progesterone molecule mainly at positions C11(α) and C21 with predominant formation of 21-hydroxyprogesterone (deoxycortone). In addition, formation of 6β,11α-dihydroxyprogesterone was also observed. Studying the effects of the growth medium composition and temperature on progesterone conversion by A. niger N402 showed that the most intense accumulation of 21-hydroxyprogesterone occurred in minimal synthetic medium at 28°C. Increasing the cultivation temperature to 37°C resulted in almost complete inhibition of the hydroxylase activity in the minimal medium. In the complete medium, a similar increase in temperature inhibited 11α-hydroxylase activity and completely suppressed 6β-hydroxylase activity, but it produced no effect on 21-hydroxylating activity.
Sigma-Aldrich
1,3-Propylene sulfite, 99%