Merck
  • Infected erythrocytes expressing DC13 PfEMP1 differ from recombinant proteins in EPCR-binding function.
Infected erythrocytes expressing DC13 PfEMP1 differ from recombinant proteins in EPCR-binding function.
Proceedings of the National Academy of Sciences of the United States of America (2018-01-18)
Yvonne Azasi, Gabriella Lindergard, Ashfaq Ghumra, Jianbing Mu, Louis H Miller, J Alexandra Rowe
ABSTRACT
Recent advances have identified a new paradigm for cerebral malaria pathogenesis in which endothelial protein C receptor (EPCR) is a major host receptor for sequestration of
Sigma-Aldrich
Bovine Serum Albumin solution, 30% in saline, IgG free, aseptically filled
Sigma-Aldrich
Fluoromount Aqueous Mounting Medium, for use with fluorescent dye-stained tissues
Sigma-Aldrich
MISSION® esiRNA, targeting human PROCR
Sigma-Aldrich
Dulbecco′s Modified Eagle′s Medium/Nutrient Mixture F-12 Ham, With L-glutamine and 15 mM HEPES, without sodium bicarbonate, powder, suitable for cell culture