Merck
  • Calcium flux between the endoplasmic reticulum and mitochondrion contributes to poliovirus-induced apoptosis.
Calcium flux between the endoplasmic reticulum and mitochondrion contributes to poliovirus-induced apoptosis.
Journal of virology (2010-09-24)
Cynthia Brisac, François Téoulé, Arnaud Autret, Isabelle Pelletier, Florence Colbère-Garapin, Catherine Brenner, Christophe Lemaire, Bruno Blondel
ABSTRACT
We show that poliovirus (PV) infection induces an increase in cytosolic calcium (Ca(2+)) concentration in neuroblastoma IMR5 cells, at least partly through Ca(2+) release from the endoplasmic reticulum lumen via the inositol 1,4,5-triphosphate receptor (IP(3)R) and ryanodine receptor (RyR) channels. This leads to Ca(2+) accumulation in mitochondria through the mitochondrial Ca(2+) uniporter and the voltage-dependent anion channel (VDAC). This increase in mitochondrial Ca(2+) concentration in PV-infected cells leads to mitochondrial dysfunction and apoptosis.
Sigma-Aldrich
Thapsigargin, ≥98% (HPLC), solid film
Sigma-Aldrich
2-Aminoethyl diphenylborinate, 97%
Sigma-Aldrich
Dantrolene, Sodium Salt, A cell-permeable blocker of intracellular calcium release from the sarcoplasmic reticulum that acts by binding to the ryanodine receptor.
Millipore
BAPTA/AM, Membrane-permeable form of BAPTA.