Merck
  • Fabrication of 8-aminocaprylic acid doped UIO-66 as sensitive solid-phase microextraction fiber for nitrosamines.
Fabrication of 8-aminocaprylic acid doped UIO-66 as sensitive solid-phase microextraction fiber for nitrosamines.
Talanta (2017-11-16)
Zhoubing Huang, Shuqin Liu, Jiangqiao Xu, Li Yin, Fen Sun, Ningbo Zhou, Gangfeng Ouyang
ABSTRACT
The introduction of functional groups into metal organic frameworks has great potential for enhancement in adsorption performance of nitrosamines using solid phase microextraction (SPME). However, pre-functionalization and post-modification generally suffer from the same limitation that the additional functional groups occupy the free volume and thereby decreasing pore volume and special surface area. Herein, we use a modulator-induced defect-formation strategy to prepare the 8-aminocaprylic acid doped UIO-66 (Am-UIO-66) with high adsorption capacity of nitrosamines. Then, the as-synthesized UIO-66 and Am-UIO-66 were fabricated as SPME fibers exhibiting good adsorption capacity towards nitrosoamines. The reason was that the 8-aminocaprylic acid creates structural defects and additional pore spaces of Am-UIO-66, thus increases the specific surface area as well as pore volume. Finally, the Am-UIO-66-coated SPME fiber was successfully applied to the determination of nitrosamines migration from latex gloves and achieved good linearity (20-2000ngL
Product Description
Supelco
SPME Fiber Assembly Kit, Assortment, StableFlex (1 cm), needle size 23 ga, Autosampler, pk of 4
Supelco
SPME Fiber Assembly Kit, Assortment #2, Fused Silica (1 cm), needle size 23 ga, Autosampler, pk of 3
Supelco
SPME Fiber Assembly Kit, Assortment #4, Fused Silica (1 cm), needle size 23 ga, Autosampler, pk of 3
Supelco
SPME Fiber Assembly Kit, Assortment #1, Fused Silica (1 cm), needle size 23 ga, Autosampler, pk of 3
Supelco
SPME Fiber Assembly Kit, Assortment #1, Fused Silica (1 cm), needle size 24 ga, Manual Holder, pk of 3
Supelco
SPME Fiber Assembly Kit, Assortment #4, Fused Silica (1 cm), size 24 ga, Autosampler, pk of 3
Supelco
SPME Fiber Assembly Kit, Assortment, StableFlex (1 cm), needle size 24 ga, Autosampler, pk of 4
Supelco
SPME Fiber Assembly Kit, Assortment #1, Fused Silica (1 cm), needle size 24 ga, Autosampler, pk of 3
Supelco
SPME Fiber Assembly Kit, Assortment #2, Fused Silica (1 cm), size 24 ga, Autosampler, pk of 3
Supelco
SPME Fiber Assembly Kit, Assortment, StableFlex (1 cm), needle size 24 ga, Manual Holder, pk of 4
Supelco
SPME Fiber Assembly Kit, Assortment #4, Fused Silica (1 cm), size 24 ga, Manual Holder, pk of 3
Supelco
SPME Fiber Assembly Kit, Assortment #2, Fused Silica (1 cm), needle size 24 ga, Manual Holder, pk of 3
Supelco
SPME Fiber Assembly Kit, Assortment #5, Fused Silica (1 cm), needle size 23 ga, Autosampler, pk of 4
Supelco
SPME Fiber Assembly, 65 μm PDMS/DVB, Metal Alloy (1 cm), needle size 23 ga, Autosampler, pk of 1, pink hub
Supelco
SPME Fiber Assembly, 85 μm CAR/PDMS, Metal Alloy (1 cm), needle size 23 ga, Autosampler, pk of 1, light blue hub
Supelco
SPME Fiber Assembly, 50/30µm DVB/CAR/PDMS, Metal Alloy (2cm), needle size 23 ga, Autosampler, pk of 1, plain gray hub
Supelco
SPME Fiber Assembly, 50/30µm DVB/CAR/PDMS, Metal Alloy (1cm), needle size 23 ga, Autosampler, pk of, plain gray hub
Supelco
SPME Fiber Assembly, 60 μm PEG, Metal Alloy (1 cm), needle size 23 ga, Autosampler, pk of 3, purple hub
Supelco
SPME Fiber Assembly, 60 μm PEG, Metal Alloy (1 cm), needle size 23 ga, Manual Holder, pk of 3, purple hub
Supelco
SPME Fiber Assembly, 85 μm CAR/PDMS, StableFlex (1 cm), needle size 24 ga, Autosampler, pk of 3, light blue hub
Supelco
SPME Fiber Assembly, 50/30µm DVB/CAR/PDMS, StableFlex (1cm), needle size 24 ga, Manual Holder, pk of 3, plain gray hub
Supelco
SPME Fiber Assembly, 100 μm PDMS, Fused Silica (1 cm), needle size 23 ga, Manual Holder, pk of 3, red hub
Supelco
SPME Fiber Assembly, 65 μm PDMS/DVB, StableFlex (1 cm), needle size 23 ga, Autosampler, pk of 3, pink hub
Supelco
SPME Fiber Assembly, 65 μm PDMS/DVB, Fused Silica (1 cm), needle size 23 ga, Autosampler, pk of 3, blue hub
Supelco
SPME Fiber Assembly, 50/30µm DVB/CAR/PDMS, StableFlex (2cm), needle size 23 ga, Autosampler, pk of 3, notched gray hub
Supelco
SPME Fiber Assembly, 85 μm Polyacrylate, Fused Silica (1 cm), needle size 23 ga, Autosampler, pk of 3, white hub
Supelco
SPME Fiber Assembly, 7 μm PDMS, Fused Silica (1 cm), needle size 23 ga, Autosampler, pk of 3, green hub
Supelco
SPME Fiber Assembly, 50/30µm DVB/CAR/PDMS, StableFlex (1 cm), needle size 23 ga, Autosampler, pk of 3, plain gray hub
Supelco
SPME Fiber Assembly, 85 μm CAR/PDMS, StableFlex (1 cm), needle size 23 ga, Autosampler, pk of 3, light blue hub
Supelco
SPME Fiber Assembly, 100 μm PDMS, Fused Silica (1 cm), needle size 24 ga, Autosampler, pk of 3, red hub