Merck
  • An insect multiligand recognition protein functions as an opsonin for the phagocytosis of microorganisms.
An insect multiligand recognition protein functions as an opsonin for the phagocytosis of microorganisms.
The Journal of biological chemistry (2010-06-04)
Chong Han Kim, Yong Pyo Shin, Mi Young Noh, Yong Hun Jo, Yeon Soo Han, Yeon Sun Seong, In Hee Lee
ABSTRACT
We characterize a novel pathogen recognition protein obtained from the lepidopteran Galleria mellonella. This protein recognizes Escherichia coli, Micrococcus luteus, and Candida albicans via specific binding to lipopolysaccharides, lipoteichoic acid, and beta-1,3-glucan, respectively. As a multiligand receptor capable of coping with a broad variety of invading pathogens, it is constitutively produced in the fat body, midgut, and integument but not in the hemocytes and is secreted into the hemolymph. The protein was confirmed to be relevant to cellular immune response and to further function as an opsonin that promotes the uptake of invading microorganisms into hemocytes. Our data reveal that the mechanism by which a multiligand receptor recognizes microorganisms contributes substantially to their phagocytosis by hemocytes. A better understanding of an opsonin with the required repertoire for detecting diverse invaders might provide us with critical insights into the mechanisms underlying insect phagocytosis.
Sigma-Aldrich
Laminarin from Laminaria digitata, polysaccharide substrate for laminarinase
Sigma-Aldrich
Curdlan from Alcaligenes faecalis
Sigma-Aldrich
Lipopolysaccharides from Escherichia coli O111:B4, purified by phenol extraction