Merck
HPLC Analysis of Aminopterin and Related Compounds on Ascentis® Express C18
HPLC Analysis of Aminopterin and Related Compounds on Ascentis® Express C18 application for HPLC
Materials
related product
Product No.
Description
Pricing
Folic acid
≥97%
Methotrexate
meets USP testing specifications
Ascentis® Express C18 (2.7 μm) HPLC Column
L × I.D. 15 cm × 4.6 mm, pk of 1 ea
CONDITIONS
column
Ascentis Express C18, 15 cm x 4.6 mm I.D., 2.7 μm particles (53829-U)
mobile phase
[A] 10 mM ammonium formate, pH 3.0; [B] acetonitrile; (90:10, A:B)
flow rate
1.0 mL/min
column temp.
35 °C
detector
UV, 280 nm
injection
5 μL
Description
Analysis Note
This application demonstrates the suitability of the Ascentis C18 for the analysis of aminopterin, folic acid and methotrexate. Structures along with the optimized chromatogram obtained on the Ascentis Express C18 are presented
Legal Information
Ascentis is a registered trademark of Merck KGaA, Darmstadt, Germany