Merck
HPLC Analysis of Ginsenosides on Ascentis® Express C18
HPLC Analysis of Ginsenosides on Ascentis® Express C18 application for HPLC
Materials
related product
Product No.
Description
Pricing
Ascentis® Express C18 (2.7 μm) HPLC Column
L × I.D. 15 cm × 4.6 mm, pk of 1 ea
CONDITIONS
column
Ascentis Express C18, 15 cm x 4.6 mm I.D., 2.7 μm particles (53829-U)
mobile phase
[A] water; [B] acetonitrile
gradient
25% B for 1.5 min; 25 to 85% B in 12 min; hold at 85% B for 1 min
flow rate
1 mL/min
column temp.
35 °C
detector
UV, 254 nm
injection
5 μL
sample
50 μg/mL in 82:18, water:methanol
Description
Legal Information
Ascentis is a registered trademark of Merck KGaA, Darmstadt, Germany