Merck
UHPLC/MS Analysis of Steroids on Titan C18
UHPLC/MS Analysis of Steroids on Titan C18 application for UHPLC
Materials
related product
Product No.
Description
Pricing
Testosterone
≥98%
Prednisolone
≥99%
Corticosterone
≥92%
Deoxycorticosterone acetate
Progesterone
≥99%
Cortisone
≥95%
CONDITIONS
column
Titan C18, 10 cm x 2.1 mm I.D., 1.9 μm particles (577124-U)
mobile phase
(A) water; (B) acetonitrile, both with 0.1% formic acid
gradient
40%B for 1 min, to 90%B in 0.5 min, held for 2 min
flow rate
0.4 mL/min
pressure
4500 psi (310 bar)
detector
MS, ESI(+), MRM
injection
2.0 μL
Description
Legal Information
null