GC Analysis of C5-C12 n-Alkanes and BTEX on SUPELCOWAX® 10
Conditions | |
|---|---|
| column | SUPELCOWAX 10, 30 m x 0.25 mm I.D., 0.25 µM (24079) |
| oven | 65 °C |
| carrier gas | helium, 30 cm/sec |
| sample | neat mixture of C5-C12 n-alkanes + BTEX, equal volumes |
| injection | wet needle, 200:1 split |
| liner | 4 mm I.D. FocusLiner™ inlet liner with taper |
| inj. temp. | 250 °C |
| detector | FID, 265 °C |
Description | |
|---|---|
| Categories | Analytical Chromatography, GC Applications, Aromatics, Hydrocarbons |
| Featured Industry | Chemicals and Industrial Polymers Petroleum |
| Legal Information | FocusLiner is a trademark of SGE Analytical Science Pty Ltd., SUPELCOWAX is a registered trademark of Sigma-Aldrich Co. LLC |
| Other Notes | Complementary products: Syringes Vials Gas Purification and Gas Management Inlet Liners, Glass Wool, and Specialized Hand Tools Septa and Specialized Hand Tools Column Ferrules, Nuts, and Specialized Hand Tools |
| suitability | application for GC |
Materials
Sorry, an unexpected error has occurred
Response not successful: Received status code 500