Merck
Advance Your Primate Studies
MILLIPLEX® Non-Human Primate Cytokine / Chemokine / Growth Factor Panel A
Cytokine and growth factor research plays a significant role in achieving a deeper understanding of the immune system and combating related diseases. As the most closely related species to humans, non-human primates are critical models for those studies. The MILLIPLEX® Non-Human Primate Cytokine/Chemokine/Growth Factor Panel A offers measurement of 48 individual immune markers, including new analytes previously not available for non-human primates. Our new 48-plex panels offer you the flexibility to choose any combination of analytes or select a 48-plex or 38-plex premixed panel. Read more about MILLIPLEX® Non-Human Primate Cytokine/Chemokine/Growth Factor Panel A.
Stay up to date on the latest immunoassay research trends and our newest product offerings by subscribing to our Analyte Update newsletter or explore our current immunoassay offerings in the Analyte Quarterly.
Endogenous levels of six analytes were tested in healthy serum/plasma samples using MILLIPLEX® Non-Human Primate Cytokine/Chemokine/Growth Factor Panel A from five non-human primate species, including rhesus macaques, cynomolgus macaques, chimpanzee, baboon, and African green monkeys.
Figure 1.Endogenous levels were tested in healthy serum/plasma samples from five non-human primate species (obtained from BioIVT), including rhesus macaques (Macaca mulatta, n=14), cynomolgus macaques (Macaca fascicularis, n=12), chimpanzee (n=9), baboon (n=9), and African green monkeys (Chlorocebus sabaeus, n=9), using the Cat. No. PRCYTA-40K panel. Concentrations of six analytes in the non-human primate species are shown.
MILLIPLEX® Non-Human Primate Cytokine/Chemokine/Growth Factor Panel A Details
Choose any combination of analytes, customize your premix, or select a 38-plex or 48-plex premixed kit.
  • Analytes: BCA-1, sCD137, sCD40L, Eotaxin, sFASL, FGF-2, Fractalkine, G-CSF, GM-CSF, Granzyme A, Granzyme B, IFNα2, IFNγ, IL-1α, IL-1β, IL-1RA, IL-2, IL-4, IL-5, IL-6, IL-7, IL-8, IL-10, IL-12 (p70), IL-15, IL-16, IL-17A, IL-17E, IL-18, IL-21, IL-22, IL-23, IL-28A, IL-31, IL-33, IP-10, I-TAC, MCP-1, MIG, MIP-1α, MIP-1β, MIP-3α, Perforin, RANTES, TGFα, TNFα, TNFβ, VEGF-A
  • Sample Type: serum, plasma, cell/tissue culture supernatants
  • Standard Curves: Broad, fixed ranges for lot-to-lot consistency, optimized for each analyte
Request Info on MILLIPLEX® Non-Human Primate Cytokine/Chemokine/Growth Factor Panel A
Experience Quality with MILLIPLEX® Assays
Our MILLIPLEX® portfolio offers the broadest selection of analytes across a wide range of disease states and species. Once the analytes of interest have been identified, you can rely on the quality that we build into each kit to produce results you can trust.
Each MILLIPLEX® panel and kit includes:
  • Quality controls (QCs) provided to qualify assay performance
  • Comparison of standard (calibrator) and QC lots to a reference lot to ensure lot-to-lot consistency
  • Optimized serum matrix to mimic native analyte environment
  • Detection antibody cocktails designed to yield consistent analyte profiles within a panel
Related Product Resources