Skip to Content
Merck

171609

Anti-β-Amyloid₄₂ (FCA3542) Rabbit pAb

Sign In to View Organizational & Contract Pricing.

Select a Size

50 μL

$825.00

$825.00


Check Cart for Availability

Request a Bulk Order

About This Item

NACRES:
NA.41
UNSPSC Code:
51111800
Species reactivity:
(Microcebus murinus, canine, human, mouse)
Species reactivity:
-
Application:
Citations:
-

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

biological source

rabbit

form

liquid

species reactivity

(Microcebus murinus, canine, human, mouse)

storage condition

OK to freeze

dilution

(ELISA
Immunoblotting (1:200-1:500)
Immunofluorescence (1:500)
Immunoprecipitation (5 µg/mL lysate)
Paraffin Sections (1:500))

isotype

IgG

storage temp.

−70°C

Quality Level

General description

Protein A purified rabbit polyclonal antibody. Recognizes Aβ1-42 and p3-related fragments of APP.

Immunogen

Human

Application

ELISA Immunoblotting (1:200-1:500)Immunofluorescence (1:500)Immunoprecipitation (5 µg/mL lysate)Paraffin Sections (1:500)

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service