Merck
Pop-Bio Products
Product photograph of two instruments stacked one on top of the other.
Pop-Bio Vü Imaging Systems for chemiluminescence and fluorescence gel documentation provide stunning images at high resolution. Move away from traditional CCD camera-based systems to the latest technology for gel and blot imaging. Simply select the tray size for your gel or blot, place in the marked position, and push in the tray. Vü immediately adapts to the gel or blot size, with images in a matter of seconds to minutes.
  • The Pop-Bio Vü-C Chemiluminescence System delivers ultra-high resolution for superb Western blot images.
  • The Pop-Bio Vü-F Fluorescence System automatically detects your sample type, capturing images using UV, blue, or white light illumination for easy imaging.
Products
Showing 1-20 of 22
Change View
Showing 1-20 of 22
Compare
Product Number
Product Name
Product Description
Pricing
You have viewed 1-20 of 22 results
Page 1 of 2