Merck
Detergents for Use in Bioprocessing
Scientist and the Bioreactor with the detergent inactivating the virus by disrupting the lipid membrane of the virus.
Detergents, such as zwitterionic or non-ionic surfactants, are used in biomanufacturing processes for a variety of applications including viral inactivation and cell lysis.
Ensuring viral safety is essential for maintaining the safety profile of drug products for patients. Unlike methods such as chromatographic separation or virus filtration that physically remove viruses, viral inactivation techniques utilize chemicals or low pH conditions to effectively reduce the presence of infectious enveloped viruses. The specific approach to viral inactivation varies based on the process and the type of molecule involved.
However, the fundamental requirement remains consistent: the use of high-quality chemicals that are free from viral contaminants and suitable for biologics manufacturing. 
What makes our detergents for biopharma Manufacturing unique?
  • Commitment to Quality: Our products are manufactured according to stringent quality standards and backed by a global Quality Management System, supporting consistency and reliability in your bioprocess.
  • Risk Mitigation: Comprehensive technical, regulatory and supply information as part of our Emprove® Program to accelerate your qualification, risk assessment, and process optimization efforts.
  • Expert Support: Technical and regulatory support to help you overcome challenges and expedite your journey to market.
Looking for a Triton™ X-100 Alternative?
Triton™ X-100 (4-tert-octylphenol polyethoxylate), a non-ionic detergent, is commonly used for virus inactivation and cell lysis as it disrupts the lipid membrane of cells and enveloped viruses. Triton™ X-100 is the main detergent templated in biomanufacturing processes, but as of January 2021, its unauthorized use has been prohibited in the European Union by the European Commission, due to its listing to REACH (Registration, Evaluation, Authorization and Restriction of Chemicals). In response to the global implications of restrictions on Triton™ X-100 detergent, we have developed the Deviron® portfolio of sustainable detergents.
The Deviron® Detergent Portfolio as a Sustainable Triton™ X-100 Alternative
Our Deviron® portfolio of detergents is a best-in-class alternative to Triton™ X-100 detergent used for biomanufacturing applications. We have developed our Deviron® detergents to address the need for alternatives to Triton™ X-100 that do not require a REACH authorization.
Key Benefits of Deviron® Detergents
  • Biodegradability (REACH* Compliance) and lack of harmful degradation products
  • Viral inactivation (5 Log Reduction Value)
  • Cell lysis efficiency
  • Minimal impact on product functionality or quality
  • Lack of process interference
  • Effective removal by downstream operations
  • Availability of sensitive detection method
  • Manufacturability of high volumes as IPEC-PQG GMP quality material
  • Impurity profile compatible with parenteral usage
*Registration, Evaluation, Authorisation and Restriction of Chemicals
DEVIRON® C16 Emprove® Evolve
Provides detailed and transparent supply chain information and documentation to support risk assessments for critical raw materials used in manufacturing processes. Non-GMP but Manufactured utilizing GMP concepts and elements; ISO9001 or equivalent.
DEVIRON® 13-S9 Emprove® Expert
Addresses higher risk applications, where the lowest microbiological and endotoxin levels are of utmost importance. Released with IPEC-PQG GMP.
Products
Showing 1-3 of 3
Change View
Showing 1-3 of 3
Compare
Product Number
Product Name
Product Description
Pricing
You have viewed 1-3 of 3 results
Page 1 of 1