Skip to Content
Merck

330740L

Avanti

SPLASH Booster

Avanti Research - A Croda Brand

Sign In to View Organizational & Contract Pricing

Select a Size


About This Item

NACRES:
NA.21
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

biological source

synthetic

form

(1:1 DCM:Methanol Solution)

manufacturer/tradename

Avanti Research - A Croda Brand

packaging

pkg of pkg of 1 x 1.1 mL (330740L-1EA)

shipped in

dry ice

storage temp.

−20°C

General description

SPLASH Booster was developed to enhance the Ultimate Splash ONE mixture, but it can also be effectively combined with any mixture. It includes five additional lipid subclasses, providing a diverse array of components that enables the most comprehensive profiling of general lipidomics samples.

Application

lipidomics

Other Notes

The SPLASH Booster Mass Spec Standard is supplied in 1.1 mL of a 1:1 DCM:methanol solution and contains the following components: 15:0-18:1-d7-PA, C13-dihydroceramide-d7(d18:0-d7/13:0), C15 Glucosyl(β) Ceramide-d7 (d18:1-d7/15:0), C15 Lactosyl(β) Ceramide-d7 (d18:1-d7/15:0), and oleic acid-d9.

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC
SPLASH is a trademark of Avanti Polar Lipids, LLC

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service