Skip to Content
Merck

860799O

Avanti

CP-LC-0729

Avanti Research - A Croda Brand

Synonym(s):

2-hexyldecyl 3-((4-((2-(dimethylamino)ethyl)amino)-3-(2-hexyldecanamido)-4-oxobutyl)thio)propanoate

Sign In to View Organizational & Contract Pricing

Select a Size

Pricing and availability is not currently available.

About This Item

Empirical Formula (Hill Notation):
C43H85N3O4S
Molecular Weight:
740.22
NACRES:
NA.21

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

biological source

synthetic

Assay

>99% (TLC)

form

powder

manufacturer/tradename

Avanti Research - A Croda Brand

packaging

pkg of pkg of 1 x 5 mg (860799O-5MG)

lipid type

transfection
cationic lipids

shipped in

dry ice

storage temp.

−20°C

Application

advanced drug delivery

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service