Skip to Content
Merck

880187P

Avanti

Cyanine 3 DBCO

new

Avanti Research - A Croda Brand

Synonym(s):

Cyanine3-N-(dibenzocyclooctyne)

Sign In to View Organizational & Contract Pricing

Select a Size


About This Item

Empirical Formula (Hill Notation):
C15H57ClN4O2
Molecular Weight:
361.09
NACRES:
NA.21
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

biological source

synthetic

Assay

>99% (TLC)

form

powder

manufacturer/tradename

Avanti Research - A Croda Brand

lipid type


fluorescent lipids

storage temp.

-10 to -25°C

SMILES string

[Cl-].CN1\C(=C\C=C\C2=[N+](CCCCCC(=O)NCCCCCC(=O)N3Cc4ccccc4C#Cc4ccccc34)c3ccccc3C2(C)C)C(C)(C)c2ccccc12

General description

Cyanine 3 DBCO is a cyanine dye functionalized with a N-(dibenzocyclooctyne) group that allows it to be used to label biomolecules such as peptides.

Application

fluorescence, flow cytometry

Features and Benefits

The DBCO terminal group offers the ability for this probe to be “clickable” and can be conjugated to a wide variety of molecules using click chemistry.

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service