Merck
Skip To
99100104
Opentrons® 50μL tip rack, filtered
Tips for AAW™ Workstation
Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
Size/SKUAvailabilityPrice
20 each
Please contact Customer Service for Availability
€321.00
About This Item
UNSPSC Code:
41121608
NACRES:
NB.02
Volume:
50 μL
Sterility:
sterile
Feature:
DNA free (Human), PCR clean, RNase and DNase free, endotoxin free, pyrogen free
Packaging:
pkg of 20 packs of 96 each
Manufacturer/tradename:
991-00104
€321.00
Please contact Customer Service for Availability
​
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
material
polypropylene
sterility
sterile
product line
AAW
feature
DNA free (Human), PCR clean, RNase and DNase free, endotoxin free, pyrogen free
manufacturer/tradename
991-00104
packaging
pkg of 20 packs of 96 each
volume
50 μL
General description
50 µL filtered pipette tips for the Opentrons Flex, provided sterile in racks or blister packs (refills). Tips are clear, non-conducting polypropylene tips, free of human DNA, PCR inhibitors, RNase/DNase, pyrogens, and endotoxins. Opentrons Flex Tips and Racks are autoclavable.
Legal Information
AAW is a trademark of Millipore Corporation
Millipore is a registered trademark of Merck KGaA, Darmstadt, Germany
Opentrons is a registered trademark of Opentrons Labworks, Inc.
Compare Similar Items
View Full Comparison
Show Differences
1 of 1
This Item
991001019910010599100110
description
Tips for AAW™ Workstation
description
Tips for AAW™ Workstation
description
Tips for AAW™ Workstation
description
Tips for AAW™ Workstation
manufacturer/tradename
991-00104
manufacturer/tradename
991-00101
manufacturer/tradename
991-00105
manufacturer/tradename
991-00110
feature
endotoxin free, PCR clean, pyrogen free, RNase and DNase free, DNA free (Human)
feature
endotoxin free, pyrogen free, DNA free (Human), RNase and DNase free, PCR clean
feature
endotoxin free, DNA free (Human), RNase and DNase free, PCR clean, pyrogen free, autoclavable
feature
endotoxin free, DNA free (Human), RNase and DNase free, autoclavable, PCR clean, pyrogen free
volume
50 μL
volume
50 μL
volume
200 μL
volume
50 μL
sterility
sterile
sterility
sterile
sterility
sterile
sterility
sterile
material
polypropylene
material
polypropylene
material
polypropylene
material
polypropylene
packaging
pkg of 20 packs of 96 each
packaging
pkg of 20 packs of 96 each
packaging
pkg of 20 packs of 96 each
packaging
pkg of 20 packs of 96 each
Choose from one of the most recent versions:
Certificates of Analysis (COA)