Skip to Content
Merck

ZRB1743

Anti-OR14I1 Antibody, clone 1C19 ZooMAb® Rabbit Monoclonal

greener alternative

recombinant, expressed in HEK 293 cells


Sign In to View Organizational & Contract Pricing.

Select a Size

Pricing and availability is not currently available.

About This Item

NACRES:
NA.43
UNSPSC Code:
12352203
Conjugate:
unconjugated
Clone:
1C19, recombinant monoclonal
Application:
ICC, WB, affinity binding assay
Citations:
-

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

biological source

rabbit

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

1C19, recombinant monoclonal

description

recombinant, expressed in HEK 293 cells

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 35.18 kDa, observed mol wt ~40 kDa

purified by

using Protein A

species reactivity

rat, human, mouse

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

affinity binding assay: suitable, immunocytochemistry: suitable, western blot: suitable

isotype

IgG

epitope sequence

N-terminal extracellular domain

Protein ID accession no.

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

Quality Level

Gene Information

human ... OR14I1(401994)

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
ZRB1260ZRB1794ZRB1056
species reactivity

rat, human, mouse

species reactivity

human, mouse

species reactivity

mouse, human, monkey

species reactivity

human

clone

1C19, recombinant monoclonal

clone

1M19, recombinant monoclonal

clone

1E19, recombinant monoclonal

clone

1D19, recombinant monoclonal

biological source

rabbit

biological source

rabbit

biological source

rabbit

biological source

rabbit

antibody form

purified antibody

antibody form

purified antibody

antibody form

purified antibody

antibody form

purified antibody

Gene Information

human ... OR14I1(401994)

Gene Information

-

Gene Information

human ... USP7(7874)

Gene Information

-

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

General description

Olfactory receptor 14I1 (UniProt: A6ND48; also known as Olfactory receptor 5BU1) is encoded by the OR14I1 (also known as OR5BU1, OR5BU1P) gene (Gene ID: 401994) in human. The olfactory receptor proteins are members of a large family of G-protein-coupled receptors (GPCR) arising from single coding-exon genes. They interact with odorant molecules in the nose to initiate a neuronal response that triggers the perception of a smell. Olfactory receptor 14I1, a part of the type II group of olfactory receptors, is a multi-pass membrane protein with four extracellular domains, seven transmembrane domains, and four cytoplasmic domains. It initiates a cascade of cellular signal transduction events that involves adenylate cyclase and protein kinase A. OR14I1 is also reported to be a host receptor a pentameric complex (gH-gL-UL128-UL130-UL131A) of human cytomegalovirus (HCMV). It is required for HCMV attachment, entry, and infection of epithelial cells. Exogenous OR14I1 protein in membrane vesicles or a synthetic peptide from the N-terminal region of OR14I1 is shown to block HCMV entry into epithelial cells. This ZooMAbZooMAb® recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals. (Ref.: Xiaofei E., et al. (2019). Proc. Natl. Acad. Sci. USA. 116(14); 7043-7052).
We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies.Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Immunogen

KLH-conjugated linear peptide corresponding to 16 amino acids from the N-terminal, extracellular domain of human Olfactory receptor 14I1 (OR14I1).

Application

Quality Control Testing

Evaluated by Western Blotting in Rat brain tissue lysate.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected OR14I1 in Rat brain tissue lysate.

Tested applications

Western Blotting Analysis: A 1:1,000 dilution from a representative lot detected OR14I1 in lysates from Mouse brain, Human brain, and SH-SY5Y cells.

Affinity Binding Assay: A representative lot of this antibody bound OR14I1 with a KD of 1.6 x 10-8 in an affinity binding assay.

Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected OR14I1 in HT1080 cells.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user

Biochem/physiol Actions

Clone 1C19 is a ZooMAb® Rabbit recombinant monoclonal antibody that specifically detects Olfactory receptor 14I1 (OR14I1). It targets an epitope within 16 amino acids from the N-terminal, extracellular domain.

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS, 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 μL.

Preparation Note

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 μL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.
Reconstitute lyophilized antibody pellet with 25μL of ultrapure water or Phosphate Buffered Saline (PBS). Please refer to our reconstitution protocol and the specific application guidance on the suggested starting dilutions and sample type.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

13 - Non Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service