Merck
The fate of ritonavir in the presence of darunavir.
International journal of pharmaceutics (2014-09-03)
D N Nguyen, G Van den Mooter
ABSTRACT
This study was the first investigation into the potential of a fixed dose combination of ritonavir and darunavir in the form of dispersible powders prepared by spray drying. A common polymer (hydroxypropyl methylcellulose, polyvinylpyrrolidone, and polyvinylpyrrolidone-vinyl acetate 64) was formulated with either ritonavir or darunavir or a combination of ritonavir and darunavir. The influence of these polymers on the supersaturation level of ritonavir and darunavir was investigated. The concentration levels of ritonavir and darunavir during these tests dropped instantly to a plateau which could be considered as amorphous solubility. Besides, the presence of darunavir always decreased the supersaturation level of ritonavir and vice versa no matter which polymers were used. Moreover, the rate and extent of release of both ritonavir and darunavir from ternary spray-dried powders were less than the releases from binary spray-dried powders. Intermolecular interaction between ritonavir and darunavir was ruled out by (1)H NMR study which means that the decrease in supersaturation level or release must be at least partially attributed to the mediated solvent process. In order to restrict the mutual influence between darunavir and ritonavir, a complex of both ritonavir and darunavir with (2-hydroxypropyl)-β-cyclodextrin was prepared and improved the dissolution rate of both ritonavir and darunavir.
Product Description
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Darunavir, ≥98% (HPLC)
Supelco
Residual Solvent - Acetonitrile(solution in DMSO), Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Sodium phosphate monobasic-16O4, 99.9 atom % 16O
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Supelco
Acetonitrile(Neat), Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Vinyl acetate, contains 3-20 ppm hydroquinone as inhibitor, ≥99%
Supelco
Acetonitrile, analytical standard
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Sodium phosphate monobasic, purum p.a., anhydrous, ≥99.0% (T)
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Supelco
Dichloromethane, Selectophore, ≥99.5%
Sigma-Aldrich
Dichloromethane, suitable for HPLC, ≥99.9%, contains 40-150 ppm amylene as stabilizer
Supelco
Dichloromethane, analytical standard
Supelco
Dimethyl sulfoxide, analytical standard
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications