Merck
  • Rapid detection of polyethylene glycol sonolysis upon functionalization of carbon nanomaterials.
Rapid detection of polyethylene glycol sonolysis upon functionalization of carbon nanomaterials.
Experimental biology and medicine (Maywood, N.J.) (2015-02-11)
Vasanth S Murali, Ruhung Wang, Carole A Mikoryak, Paul Pantano, Rockford Draper
ABSTRACT
Polyethylene glycol (PEG) and related polymers are often used in the functionalization of carbon nanomaterials in procedures that involve sonication. However, PEG is very sensitive to sonolytic degradation and PEG degradation products can be toxic to mammalian cells. Thus, it is imperative to assess potential PEG degradation to ensure that the final material does not contain undocumented contaminants that can introduce artifacts into experimental results. Described here is a simple and inexpensive polyacrylamide gel electrophoresis method to detect the sonolytic degradation of PEG. The method was used to monitor the integrity of PEG phospholipid constructs and branched chain PEGs after different sonication times. This approach not only helps detect degraded PEG, but should also facilitate rapid screening of sonication parameters to find optimal conditions that minimize PEG damage.
Product Description
Avanti
DSPE-PEG(2000) Amine, Avanti Research - A Croda Brand
Pricing and availability is not currently available.
Avanti
DSPE-PEG(2000) Amine, Avanti Research - A Croda Brand 880128C
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium phosphate, 96%
Pricing and availability is not currently available.