Merck
GC Analysis of QA Test Mix on SLB®-IL60 before/after Thermal Stress Test, Comparison to a Polyethylene Glycol (PEG) Column
GC Analysis of QA Test Mix on SLB®-IL60 before/after Thermal Stress Test, Comparison to a Polyethylene Glycol (PEG) Column suitable for GC
Materials
related product
Product No.
Description
Pricing
Polar Column Test Mix
certified reference material, 500 μg/mL each component in dichloromethane
SLB®-IL60 Capillary GC Column
L × I.D. 30 m × 0.25 mm, df 0.20 μm
CONDITIONS
column
PEG, 30 m x 0.25 mm I.D., 0.25 μm
column
SLB-IL60, 30 m x 0.25 mm I.D., 0.20 μm (29505-U)
oven
155 °C (25 min) for PEG 3; 130 °C (25 min) for SLB-IL60
inj. temp.
250 °C
detector
FID, 250 °C
carrier gas
helium, 20 cm/sec for PEG 3; helium, 25 cm/sec for SLB-IL60
injection
1.0 μL 100:1 split
liner
4 mm I.D., split/splitless type, single taper wool packed FocusLiner design
sample
polar column test mix (47302), 9 analytes, each at 500 μg/mL in methylene chloride
Description
Legal Information
FocusLiner is a trademark of SGE Analytical Science Pty Ltd.
SLB is a registered trademark of Merck KGaA, Darmstadt, Germany