LC/MS Analysis of Amphetamines on Ascentis® Express HILIC
CONDITIONS
column
Ascentis Express HILIC, 10 cm x 3 mm I.D., 2.7 μm particles (53970-U)
mobile phase
[A] 1 mM ammonium acetate in [B] 10:90 (v/v) water:acetonitrile
flow rate
0.6 mL/min
pressure
1189 psi (82 bar)
column temp.
35 °C
detector
ESI(+), full scan, m/z 110-200
injection
2 μL
sample
10 mg/L each in acetonitrile
Description
Legal Information
Ascentis is a registered trademark of Merck KGaA, Darmstadt, Germany