HomeWebinarsWebinar: ISO 16140-3:2021
Webinar: ISO 16140-3:2021
Webinar
The webinar introduces the new EN ISO 16140-3 on the verification of methods in food microbiology. This standard provides the protocols and criteria to demonstrate that a user laboratory can satisfactorily perform a validated method and that this method is fit for its intended purpose. The standard includes considerations about method and laboratory scope, number of replicates, sample inoculation as well as the evaluation and interpretation of results to confirm that the method is well implemented and aligned with the food matrices analyzed in the user laboratory.
What is covered?
  • Validation and verification
  • Microbiological method verification
    • Reference methods
    • Alternative methods
  • Protocol for qualitative method verification
  • Protocol for quantitative method verification
Speaker
David Tomás Fornés
Merck
Application Specialist
David is an application specialist at Merck. He is an expert in several ISO technical committees that develop reference methods for food microbiology.
Webinar Information
Microbiological testing
  • Bioburden testing
  • Duration:1 hour
  • Language:English
  • Presented:March 30, 2021