All Photos(1)

BR122760

BRAND® Bottle-Top Vacuum Filter Funnel

pore size 0.2 μm, sterile, polyethersulfone membrane, pack of 12

NACRES:
NB.24

material

plastic housing
polyethersulfone membrane

sterility

sterile

feature

cap
graduations

packaging

pack of 12

filter capacity

500 mL

funnel capacity

500 mL

pore size

0.2 μm

Legal Information

BRAND is a registered trademark of BRAND GMBH + CO KG

Certificate of Analysis

Certificate of Origin

Dinesh Erram et al.
Journal of medical entomology, 57(1), 25-32 (2019-10-12)
Laboratory rearing procedures of Culicoides stellifer Coquillett (Diptera: Ceratopogonidae) were evaluated with an aim towards colonization of this species. Eggs collected from field-collected gravid females were placed on 0.25% agar slants and given a diet of 1) nematodes (Panagrellus redivivus...
Yuanjun Zhao et al.
Water research, 177, 115757-115757 (2020-04-13)
Commercially available powdered activated carbon (PAC) with a median diameter of 12-42 μm was ground into 1 μm sized superfine PAC (SPAC) and 200 nm sized submicron SPAC (SSPAC) and investigated as a pretreatment material for the prevention of hydraulically irreversible membrane fouling...
Andrew C Schuerger et al.
Life (Basel, Switzerland), 10(6) (2020-05-30)
The search for life on Mars is predicated on the idea that Earth and Mars life (if present) should be both carbon- and water-based with similar forms of evolution. However, the astrobiology community can currently only investigate plausible Martian microbial...
Joo-Hee Choi et al.
International journal of medical sciences, 17(5), 632-639 (2020-03-27)
Background: Pumpkin (Curcubita sp.) is a natural product that is commonly used in folk medicine. However, the inhibitory effect and molecular mechanisms of tendril of Cucurbita Moschata Duch. (TCMD) on osteoclast differentiation have yet to be clearly elucidated. Thus, the...

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service