Merck
99900197
Opentrons Flex HEPA/UV Module
new
Opentrons Flex HEPA/UV Module
Select a Size
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
packaging
pack of 1 ea
manufacturer/tradename
999-00197
technique(s)
analytical sample preparation: suitable
application(s)
clinical testing
compatibility
Opentrons Flex Robot
General description
The OPENTRONSFLEX HEPA/UV Module enables you to run sensitive contamination-prone applications. It removes 99.99% of 0.3 μm DNA-containing particulates and biological contaminants like bacteria, fungi, and other microorganisms from the air, creating a clean work environment inside the AAW Automated Assay Workstation.
Legal Information
AAW is a trademark of Millipore Corporation
OPENTRONSFLEX is a trademark of Opentrons Labworks, Inc.