Merck
All Photos(1)

A2174

Sigma-Aldrich

L-Ascorbic acid

suitable for plant cell culture

Sign Into View Organizational & Contract Pricing

Select a Size

Synonym(s):
L-Threoascorbic acid, Antiscorbutic factor, Vitamin C
Empirical Formula (Hill Notation):
C6H8O6
CAS Number:
Molecular Weight:
176.12
Beilstein:
84272
EC Number:
MDL number:
PubChem Substance ID:

Assay

≥98% ( with iodine, titration)

form

powder

mol wt

Mw 176.12 g/mol

technique(s)

cell culture | plant: suitable

color

white to off-white

pH

1.0-2.5 (25 °C, 176 g/L in water)

mp

190-194 °C (dec.)

solubility

water: soluble 50 mg/mL

SMILES string

OC([C@]([C@@H](O)CO)([H])O1)=C(O)C1=O

InChI

1S/C6H8O6/c7-1-2(8)5-3(9)4(10)6(11)12-5/h2,5,7-10H,1H2/t2-,5+/m0/s1

InChI key

CIWBSHSKHKDKBQ-JLAZNSOCSA-N

Gene Information

human ... SLC23A2(9962)

Looking for similar products? Visit Product Comparison Guide

Caution

May darken in storage.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Documents related to the products that you have purchased in the past have been gathered in the Document Library for your convenience.

Visit the Document Library

Difficulty Finding Your Product Or Lot/Batch Number?

Product numbers are combined with Pack Sizes/Quantity when displayed on the website (example: T1503-25G). Please make sure you enter ONLY the product number in the Product Number field (example: T1503).

Example:

T1503
Product Number
-
25G
Pack Size/Quantity

Additional examples:

705578-5MG-PW

PL860-CGA/SHF-1EA

MMYOMAG-74K-13

1000309185

enter as 1.000309185)

Having trouble? Feel free to contact Technical Service for assistance.

Lot and Batch Numbers can be found on a product's label following the words 'Lot' or 'Batch'.

Aldrich Products

  • For a lot number such as TO09019TO, enter it as 09019TO (without the first two letters 'TO').

  • For a lot number with a filling-code such as 05427ES-021, enter it as 05427ES (without the filling-code '-021').

  • For a lot number with a filling-code such as STBB0728K9, enter it as STBB0728 without the filling-code 'K9'.

Not Finding What You Are Looking For?

In some cases, a COA may not be available online. If your search was unable to find the COA you can request one.

Request COA

Customers Also Viewed

Slide 1 of 10

1 of 10

L-Ascorbic acid solution 1.0 mg/mL in acetonitrile: water, certified reference material, Cerilliant®

Supelco

V-038

L-Ascorbic acid solution

(+)-Sodium L-ascorbate powder, BioReagent, suitable for cell culture

Sigma-Aldrich

A4034

(+)-Sodium L-ascorbate

L-Ascorbic acid puriss. p.a., ≥99.0% (RT)

Sigma-Aldrich

95210

L-Ascorbic acid

Ascorbic acid European Pharmacopoeia (EP) Reference Standard

A1300000

Ascorbic acid

L-Ascorbic acid analytical standard

Supelco

47863

L-Ascorbic acid

Ascorbic Acid Pharmaceutical Secondary Standard; Certified Reference Material

Supelco

PHR1008

Ascorbic Acid

Ascorbic acid British Pharmacopoeia (BP) Reference Standard

BP461

Ascorbic acid

(+)-Sodium L-ascorbate BioXtra, ≥99.0% (NT)

Sigma-Aldrich

11140

(+)-Sodium L-ascorbate

Takashi Yoshida et al.
Nature chemical biology, 2(11), 596-607 (2006-09-26)
Transient receptor potential (TRP) proteins form plasma-membrane cation channels that act as sensors for diverse cellular stimuli. Here, we report a novel activation mechanism mediated by cysteine S-nitrosylation in TRP channels. Recombinant TRPC1, TRPC4, TRPC5, TRPV1, TRPV3 and TRPV4 of
Marc P Bradshaw et al.
Critical reviews in food science and nutrition, 51(6), 479-498 (2011-09-21)
Extensive reviews of research are available on the use of ascorbic acid, and its consequent degradation pathways, in physiological conditions or food matrices. However, very little information can be found for wine-related systems. This review highlights the relevant chemistry and
Noriko Umegaki-Arao et al.
Science translational medicine, 6(264), 264ra164-264ra164 (2014-11-28)
Revertant mosaicism is a naturally occurring phenomenon involving spontaneous correction of a pathogenic gene mutation in a somatic cell. It has been observed in several genetic diseases, including epidermolysis bullosa (EB), a group of inherited skin disorders characterized by blistering
Josué Kunjom Mfopou et al.
Stem cell research, 12(1), 166-177 (2013-11-19)
Definitive endoderm (DE) differentiation from mouse embryonic stem cell (mESC) monolayer cultures has been limited by poor cell survival or low efficiency. Recently, a combination of TGFβ and Wnt activation with BMP inhibition improved DE induction in embryoid bodies cultured
Zi Yin et al.
Biomaterials, 44, 173-185 (2015-01-27)
Physical topographic cues from various substrata have been shown to exert profound effects on the growth and differentiation of stem cells due to their niche-mimicking features. However, the biological function of different topographic materials utilized as bio-scaffolds in vivo have

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service