All Photos(1)

MAK407

Sigma-Aldrich

Nitric Oxide Synthase (NOS) Activity Assay Kit (Colorimetric)

new

sufficient for 100 colorimetric tests

Quality Level

detection method

colorimetric

storage temp.

−70°C

Related Categories

General description

Nitric oxide synthases (NOS) are a family of enzymes that catalyze the production of nitric oxide (NO) from L-arginine. Nitric oxide (NO) plays an important role in neurotransmission, vascular regulation, immune response and apoptosis. In the presence of NADPH, FAD, FMN, (6R)-5,6,7,8-tetrahydrobiopterin, calmodulin and heme, NOS catalyzes a five-electron oxidation of the guanidino nitrogen of L-arginine with molecular oxygen to generate NO and L-citrulline. There are three isoforms of NOS: endothelial (eNOS), neuronal (nNOS), and inducible (iNOS). nNOS accounts for the production of NO in the central nervous system, where NO participates in cell communication and information storage. eNOS produces NO in blood vessels and is involved in regulation of vascular function. In contrast to other isoforms, iNOS is expressed de novo under oxidative stress conditions and produces large amounts of NO as part of the body′s defense mechanism.

Features and Benefits

Nitric Oxide Synthase (NOS) Activity Assay Kit:
  • Compatible with high-throughput handling systems.
  • Sensitive, detecting as low as 5 μU of NOS activity.

Suitability

The kit is suitable for the detection of NOS activity in purified recombinant protein and tissue or cell extracts.

Principle

In this assay, nitric oxide generated by NOS undergoes a series of reactions and reacts with Griess Reagent 1 and 2 to generate a colored product with a strong absorbance at 540 nm

signalword

Danger

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Acute 1 - Carc. 2 - Eye Irrit. 2 - Met. Corr. 1 - Muta. 2 - Ox. Liq. 3 - Repr. 2 - Skin Irrit. 2 - STOT RE 2

Target Organs

Nervous system

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK Germany

WGK 3

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service