Skip to Content
Merck
All Photos(1)

Key Documents

Z744223

KDS Gemini 88 Plus Dual Rate Syringe Pump, Infusion/Withdrawal

new

AC/DC input 100-240 V, UK plug

Synonym(s):

dosing pump, infusion pump, withdrawal pump

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121500
Pricing and availability is not currently available.

feature

no. of channels 2
CE, RoHS, WEEE Approved
accuracy ± 0.25%

packaging

pack of 1 ea

manufacturer/tradename

KD Scientific 788088

L × D × H

28 cm (11 in.) × 39 cm (15 in.) × 21 cm (8 in.)

volume

0.0005—60 mL

weight

9.09 kg (21 lb)

Looking for similar products? Visit Product Comparison Guide

General description

This dual rate syringe pump can infuse simultaneously at different rates, or infuse with one syringe and withdraw with the other. When combined with a valve box, it provides the continuous delivery of a peristaltic or piston pump with the accuracy, absence of pulsation and low flow rates of a syringe pump. This functionality allows continuous infusion or withdrawal, 24 hours a day, 365 days a year with the accuracy and low flow of every KD Scientific syringe pump.

Features and Benefits

  • Two separate syringe pumps in one instrument
  • Large touch screen display allows quick and easy setup
  • High or low pressure operation
  • Compatible with a wide variety of syringes
  • Continuous fluid delivery
  • TTL, USB & RS-232 Communication


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service