Merck
All Photos(1)

150705

Supelco

Sodium hydroxide solution

0.1 M, Set incl. Absorber cartridge with soda lime, for 3S adapter technology

Linear Formula:
NaOH
CAS Number:
Molecular Weight:
40.00
MDL number:

vapor pressure

3 mmHg ( 37 °C)

form

liquid

reaction suitability

reaction type: Acid-base reactions

concentration

0.1 M

technique(s)

titration: suitable

volume

1 L

SMILES string

[OH-].[Na+]

InChI

1S/Na.H2O/h;1H2/q+1;/p-1

InChI key

HEMHJVSKTPXQMS-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Contains 1506971000 (NaOH solution 0.1 M) and 1507040001 (cartridge filled with soda lime). Before opening the 3S cap, insert cartridge into cap.

Please see here SigmaAldrich.com/3S

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8A - Combustible, corrosive hazardous materials

WGK

WGK 1

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

Related Content

3S Reagents for Volumetric Titration

3S is an intelligent packaging solution protected with an originality seal. The cap provides information via Radio-frequency identification or RFID, enabling automated data transfer between the reagent and titration instrument. Intelligent communication for improved performance.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service