BR
EN
Applications
Products
Services
Support
Home
Search Results
2′,7′-Dichlorofluorescin Diacetate
(2)
2′,7′-Dichlorofluorescin Diacetate
Synonyms:
2′,7′-Dichlorodihydrofluorescein diacetate, 2′,7′-Dichlorofluorescin Diacetate
Empirical Formula (Hill Notation):
C
24
H
16
Cl
2
O
7
CAS Number:
4091-99-0
Molecular Weight:
487.29
Product Comparison Guide
Use the product attributes below to configure the comparison table. (Select up to 3 total.)
Select Attribute
Select Attribute
Select Attribute
Sort by: Default
Select Attributes
Product Number
Product Description
Pricing
D6883
2′,7′-Dichlorofluorescin diacetate, ≥97%
Expand
View Pricing
287810
2′,7′-Dichlorofluorescin Diacetate, Cell-permeable fluorogenic probe that is useful for the detection of reactive oxygen species (ROS) and nitric oxide (•NO) and for the determination of the degree of overall oxidative stress.
Expand
View Pricing