BR
EN
Applications
Products
Services
Support
Home
Search Results
Accellerase
™
1000
(1)
Accellerase
™
1000
Synonyms:
Cellulase from
Trichoderma reesei
CAS Number:
9012-54-8
Product Comparison Guide
Use the product attributes below to configure the comparison table. (Select up to 3 total.)
Select Attribute
Select Attribute
Select Attribute
Sort by: Default
Select Attributes
Product Number
Product Description
Pricing
C9870
Accellerase
™
1000
Expand
View Pricing