(±)-Epichlorohydrin analytical standard

(±)-Epichlorohydrin

(±)-Epichlorohydrin analytical standard
Synonyms:
(±)-2-(Chloromethyl)oxirane, 1-Chloro-2,3-epoxypropane
Empirical Formula (Hill Notation):
C3H5ClO
CAS Number:
Molecular Weight:
92.52
EC Number:

Product Comparison Guide

Use the product attributes below to configure the comparison table. (Select up to 3 total.)

Select Attribute
Select Attribute
Select Attribute
Sort by: Default
Product Number
Product Description
Pricing
(±)-Epichlorohydrin, purum, ≥99% (GC)
(±)-Epichlorohydrin, analytical standard