(±)-Jasmonic acid analytical standard

(±)-Jasmonic acid

(±)-Jasmonic acid analytical standard
Empirical Formula (Hill Notation):
C12H18O3
CAS Number:
Molecular Weight:
210.27

Product Comparison Guide

Use the product attributes below to configure the comparison table. (Select up to 3 total.)

Select Attribute
Select Attribute
Select Attribute
Sort by: Default
Product Number
Product Description
Pricing
(±)-Jasmonic acid, BioReagent, suitable for plant cell culture, liquid
(±)-Jasmonic acid, analytical standard