  • Search Results
  • MICAN, a new fluorophore for vital and non-vital staining of human cells
MICAN, a new fluorophore for vital and non-vital staining of human cells
Toxicology in vitro (2018)
Nagy Z, et al.