BR
EN
Applications
Products
Services
Support
Home
Search Results
Rhodamine B as a probe for phase transition in liquid solutions
Rhodamine B as a probe for phase transition in liquid solutions
arXiv (2016)
Shishkin I, et al.