  • Search Results
  • Lysozyme/EDTA disodium salt and modified-atmosphere packaging to prolong the shelf life of burrata cheese
Lysozyme/EDTA disodium salt and modified-atmosphere packaging to prolong the shelf life of burrata cheese
Journal of Dairy Science (2011)
Conte A, et al.