  • Search Results
  • Conformational landscape of the p28-bound human proteasome regulatory particle
Conformational landscape of the p28-bound human proteasome regulatory particle
Molecular Cell (2017)
Lu Y, et al.