  • Search Results
  • Novel Miniaturized Drug Conjugate Leverages HSP90-driven Tumor Accumulation to Overcome PI3K Inhibitor Delivery Challenges to Solid Tumors.
Novel Miniaturized Drug Conjugate Leverages HSP90-driven Tumor Accumulation to Overcome PI3K Inhibitor Delivery Challenges to Solid Tumors.
Molecular cancer therapeutics (2020-06-06)
Samantha Perino, Benoit Moreau, Jessica Freda, Amanda Cirello, Brian H White, James M Quinn, Kristina Kriksciukaite, Ashwajith Someshwar, Janel Romagnoli, Megan Robinson, Sara Movassaghian, Tyler Cipriani, Richard Wooster, Mark T Bilodeau, Kerry A Whalen
ABSTRACT
The PI3K pathway is considered a master regulator for cancer due to its frequent activation, making it an attractive target for pharmacologic intervention. While substantial efforts have been made to develop drugs targeting PI3K signaling, few drugs have been able to achieve the inhibition necessary for effective tumor control at tolerated doses. HSP90 is a chaperone protein that is overexpressed and activated in many tumors and as a consequence, small-molecule ligands of HSP90 are preferentially retained in tumors up to 20 times longer than in normal tissue. We hypothesize that the generation of conjugates that use a HSP90-targeting ligand and a payload such as copanlisib, may open the narrow therapeutic window of this and other PI3K inhibitors. In support of this hypothesis, we have generated a HSP90-PI3K drug conjugate, T-2143 and utilizing xenograft models, demonstrate rapid and sustained tumor accumulation of the conjugate, deep pathway inhibition, and superior efficacy than the PI3K inhibitor on its own. Selective delivery of T-2143 and the masking of the inhibitor active site was also able to mitigate a potentially dose-limiting side effect of copanlisib, hyperglycemia. These data demonstrate that by leveraging the preferential accumulation of HSP90-targeting ligands in tumors, we can selectively deliver a PI3K inhibitor leading to efficacy in multiple tumor models without hyperglycemia in mice. These data highlight a novel drug delivery strategy that allows for the potential opening of a narrow therapeutic window through specific tumor delivery of anticancer payloads and reduction of toxicity.
Product Description
Supelco
Dichlorvos, PESTANAL®, analytical standard
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Glyburide, meets USP testing specifications