  • Search Results
  • Increased Alu RNA processing in Alzheimer brains is linked to gene expression changes.
Increased Alu RNA processing in Alzheimer brains is linked to gene expression changes.
EMBO reports (2021-03-02)
Yubo Cheng, Luke Saville, Babita Gollen, Ana Alvarez Veronesi, Majid Mohajerani, Jeffrey T Joseph, Athanasios Zovoilis
ABSTRACT
Despite significant steps in our understanding of Alzheimer's disease (AD), many of the molecular processes underlying its pathogenesis remain largely unknown. Here, we focus on the role of non-coding RNAs produced by small interspersed nuclear elements (SINEs). RNAs from SINE B2 repeats in mouse and SINE Alu repeats in humans, long regarded as "junk" DNA, control gene expression by binding RNA polymerase II and suppressing transcription. They also possess self-cleaving activity that is accelerated through their interaction with certain proteins disabling this suppression. Here, we show that similar to mouse SINE RNAs, human Alu RNAs, are processed, and the processing rate is increased in brains of AD patients. This increased processing correlates with the activation of genes up-regulated in AD patients, while increased intact Alu RNA levels correlate with down-regulated gene expression in AD. In vitro assays show that processing of Alu RNAs is accelerated by HSF1. Overall, our data show that RNAs from SINE elements in the human brain show a similar pattern of deregulation during amyloid beta pathology as in mouse.
Sigma-Aldrich
Chloroform, contains 100-200 ppm amylenes as stabilizer, ≥99.5%
Sigma-Aldrich
Minimum Essential Medium Eagle, With Earle′s salts, L-glutamine and sodium bicarbonate, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Nutrient Mixture F-12 Ham, With L-glutamine and sodium bicarbonate, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Fetal Bovine Serum, USA origin, Heat Inactivated, sterile-filtered, suitable for cell culture, suitable for insect cell culture, suitable for hybridoma