Supelco
L-Glutamine, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
L-Glutamine, certified reference material, TraceCERT®
Sigma-Aldrich
L-Glutamine, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
L-Glutamine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
L-Glutamine, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
L-Glutamine, ReagentPlus®, ≥99% (HPLC)
SAFC
RPMI-1640 Medium, Hybri-Max™, Modified, with L-glutamine, 4500 mg/L glucose and 15mM HEPES, without sodium bicarbonate, powder, suitable for hybridoma
Sigma-Aldrich
RPMI-1640 Medium, Modified, with sodium bicarbonate, without methionine, cystine and L-glutamine, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
RPMI-1640 Medium, Dutch Modification, with sodium bicarbonate and 20mM HEPES, without L-glutamine, liquid, sterile-filtered, suitable for cell culture
SAFC
RPMI-1640 Medium, 10 ×, Without L-glutamine, folic acid and sodium bicarbonate, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
RPMI-1640 Medium, Modified, with 20 mM HEPES and L-glutamine, without sodium bicarbonate, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
RPMI-1640 Medium, With L-glutamine, without glucose and sodium bicarbonate, powder, suitable for cell culture
SAFC
RPMI-1640 Medium, AutoMod™, without L-glutamine and sodium bicarbonate, powder, suitable for cell culture
Sigma-Aldrich
RPMI-1640 Medium, With L-glutamine and sodium bicarbonate, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
RPMI-1640 Medium, With sodium bicarbonate, without L-glutamine, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
RPMI-1640 Medium, Modified, with sodium bicarbonate, without L-glutamine and phenol red, liquid, sterile-filtered, suitable for cell culture
SAFC
RPMI-1640 Medium, HEPES Modification, with L-glutamine and 25mM HEPES, without sodium bicarbonate, powder, suitable for cell culture
SAFC
RPMI-1640 Medium, HEPES Modification, With 25 mM HEPES, without L-glutamine., liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
RPMI-1640 Medium, With L-glutamine, without sodium bicarbonate, powder, suitable for cell culture
Sigma-Aldrich
RPMI-1640 Medium, Modified, with L-glutamine, without phenol red and sodium bicarbonate, powder, suitable for cell culture
Sigma-Aldrich
Lipopolysaccharides from Escherichia coli O111:B4, purified by ion-exchange chromatography, TLR ligand tested
Sigma-Aldrich
RPMI-1640 Medium, With L-glutamine and sodium bicarbonate. Without arginine, leucine, lysine, and phenol red, liquid, sterile-filtered, suitable for cell culture, designed for isotope labeling for cell culture applications
Sigma-Aldrich
StableCell™ RPMI-1640, With stable glutamine and sodium bicarbonate, liquid, sterile-filtered, suitable for cell culture
SAFC
RPMI-1640 Medium, with 2.05 mM L-glutamine, with 25mM HEPES, liquid, sterile-filtered, suitable for cell culture